Skip to Content
Merck
All Photos(3)

Key Documents

738891

Sigma-Aldrich

2,5-Bis(trimethylstannyl)thiophene

97%

Synonym(s):

1,1′-(2,5-thiophenediyl)bis[1,1,1-trimethyl] Stannane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20SSn2
CAS Number:
Molecular Weight:
409.75
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

93-102 °C

SMILES string

C[Sn](C)(C)c1ccc(s1)[Sn](C)(C)C

InChI

1S/C4H2S.6CH3.2Sn/c1-2-4-5-3-1;;;;;;;;/h1-2H;6*1H3;;

InChI key

KKRPPVXJVZKJON-UHFFFAOYSA-N

Application


  • Technical note: Evaluating nuclear magnetic resonance spectroscopy for determining body composition in Holstein dairy calves using deuterium oxide dilution methods.: This research explores the application of nuclear magnetic resonance spectroscopy, combined with deuterium oxide dilution, to accurately determine body composition in dairy calves. This method could offer a non-invasive alternative for assessing nutritional status and growth metrics in livestock management (Chapman et al., 2017).

  • Estimating lean mass over a wide range of body composition: a calibration of deuterium dilution in the arctic ground squirrel.: This study calibrates deuterium oxide dilution for estimating lean mass in Arctic ground squirrels. The findings could enhance our understanding of body composition changes in hibernating species, potentially influencing studies on metabolic adaptations and energy usage (Lee et al., 2011).

  • Long-day photoperiod that enhances puberty does not limit body growth in Holstein heifers.: Investigates how extending daylight hours affects the growth and reproductive timing in dairy heifers. Deuterium oxide was used to measure body water and composition, providing insights into how light exposure can influence animal development (Rius et al., 2005).



Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiyan Li et al.
Journal of the American Chemical Society, 136(41), 14589-14597 (2014-09-30)
New electron-acceptor materials are long sought to overcome the small photovoltage, high-cost, poor photochemical stability, and other limitations of fullerene-based organic photovoltaics. However, all known nonfullerene acceptors have so far shown inferior photovoltaic properties compared to fullerene benchmark [6,6]-phenyl-C60-butyric acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service