Skip to Content
Merck
All Photos(1)

Key Documents

734594

Sigma-Aldrich

N,N′-Di(1-naphthyl)-N,N′-diphenyl-(1,1′-biphenyl)-4,4′-diamine

96%

Synonym(s):

N,N′-Bis(naphthalen-1-yl)-N,N′-bis(phenyl)benzidine, NPB, NPD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H32N2
CAS Number:
Molecular Weight:
588.74
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

96%

form

solid

mp

275-280 °C
279-283 °C (lit.)

Orbital energy

HOMO 5.5 eV 
LUMO 2.4 eV 

OLED Device Performance

ITO/NPD/TCTA/BCPO:Ir(piq)3 (7-8%)/BCP/Alq3/LiF/Al

  • Color: red
  • Max. Luminance: 24529 Cd/m2
  • Max. EQE: 17 %
  • Turn-On Voltage: 2.7 V

ITO/NPD/TCTA/BCPO:Ir(ppy)3 (7-8%)/BCP/Alq3/LiF/Al
  • Color: green
  • Max. Luminance: 207839 Cd/m2
  • Max. EQE: 21.6 %
  • Turn-On Voltage: 2.1 V

ITO/NPD/mCP/BCPO:FIrpic (8%)/TAZ/LiF/Al
  • Color: blue
  • Max. Luminance: 35677 Cd/m2
  • Max. EQE: 23.5 %
  • Turn-On Voltage: 2.8 V

ITO/PEDOT:PSS/NPD/TCTA/m-CBTZ:Ir(Bt)2(acac) (10%)/TPBI/LiF/Al
  • Color: yellow
  • Max. Luminance: 106000 Cd/m2
  • Max. EQE: 17.5 %
  • Turn-On Voltage: 3 V

SMILES string

c1ccc(cc1)N(c2ccc(cc2)-c3ccc(cc3)N(c4ccccc4)c5cccc6ccccc56)c7cccc8ccccc78

InChI

1S/C44H32N2/c1-3-17-37(18-4-1)45(43-23-11-15-35-13-7-9-21-41(35)43)39-29-25-33(26-30-39)34-27-31-40(32-28-34)46(38-19-5-2-6-20-38)44-24-12-16-36-14-8-10-22-42(36)44/h1-32H

InChI key

IBHBKWKFFTZAHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hole transporting material for OLED devices

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Courtney M Wheatley et al.
Respiratory medicine, 109(8), 1001-1011 (2015-06-17)
Despite the demonstrated advantageous systemic changes in response to regular exercise for individuals with cystic fibrosis (CF), exercise is still viewed as an elective rather than a vital component of therapy, and it is likely that these benefits extend to
Shakeel S Dalal et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(14), 4227-4232 (2015-04-02)
Physical vapor deposition is commonly used to prepare organic glasses that serve as the active layers in light-emitting diodes, photovoltaics, and other devices. Recent work has shown that orienting the molecules in such organic semiconductors can significantly enhance device performance.
Highly efficient and stable organic light-emitting diode using 4,4'-bis(N-carbazolyl)-9,9'-spirobifluorene as a thermally stable host material
Tsuzuki, T.; Tokito, S.
Applied Physics Letters, 94, 033302-033302 (2009)
Phenylcarbazole-dipyridyl triazole hybrid as bipolar host material for phosphorescent OLEDs
Hung, W.; et al.
Journal of Materials Chemistry, 22, 5410-5418 (2012)
Sun-Ju Nam Goung et al.
Environmental science and pollution research international, 22(9), 7176-7182 (2014-12-17)
The aims of this study were to provide basic data for determining policies on air quality for multi-user facilities, including the legal enrollment of the indoor air quality regulation as designated by the Ministry of Environment, and to establish control

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service