Skip to Content
Merck
All Photos(3)

Key Documents

729965

Sigma-Aldrich

1H,1H,2H,2H-Perfluorododecyltrichlorosilane

97%

Synonym(s):

3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Heneicosafluorododecyltrichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)9CH2CH2SiCl3
CAS Number:
Molecular Weight:
681.57
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

50-55 °C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Si](Cl)(Cl)Cl

InChI

1S/C12H4Cl3F21Si/c13-37(14,15)2-1-3(16,17)4(18,19)5(20,21)6(22,23)7(24,25)8(26,27)9(28,29)10(30,31)11(32,33)12(34,35)36/h1-2H2

InChI key

ZFUVZJADECZZMS-UHFFFAOYSA-N

General description

1H,1H,2H,2H-Perfluorododecyltrichlorosilane(FTCS) is a fluorosilane that can be used as a low surface energy material. It forms a hydrophobic coating with high wetting properties and contact angle.

Application

1H,1H,2H,2H-perfluorodecyltrichlorosilane (FAS21, FDDTS) was used to prepare a hydrophobic surface in a study. Self assembled monolayers of FDDTS were formed on titania substrates to render the surfaces hydrophobic.
FTCS can be incorporated in polyvinylidiene fluoride(PVDF) with titanium oxide(TiO2) which can be used for membrane distillation for water treatment and anti-fouling based application.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Superhydrophobic modification of TiO2 nanocomposite PVDF membranes for applications in membrane distillation.
Razmjou A, et al.
Journal of Membrane Science , 415, 850-863 (2012)
Superhydrophobic modification of TiO 2 nanocomposite PVDF membranes for applications in membrane distillation.
Journal of Membrane Science, 415, 850-863 (2012)
Superhydrophobic membranes via facile bio-inspired mineralization for vacuum membrane distillation.
Zhong W, et al.
Journal of Membrane Science , 540, 98-107 (2017)
A paper microfluidic cartridge for automated staining of malaria parasites with an optically transparent microscopy window.
Lab on a chip, 14(12), 2040-2046 (2014)
Effect of templating agents on the properties and membrane distillation performance of TiO2-coated PVDF membranes.
Meng S, et al.
Journal of Membrane Science , 450, 48-59 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service