Skip to Content
Merck
All Photos(1)

Documents

729116

Sigma-Aldrich

Poly(ethylene glycol) methyl ether tosylate

average MN 900, tosylate, methoxy

Synonym(s):

Polyethylene glycol, Methoxy PEG tosylate, Methoxypolyethylene glycol tosylate, PEG tosylate, mPEG tosylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)nCH2CH2OSO2C6H4CH3
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene glycol) methyl ether tosylate, average Mn 900

form

solid or liquid

Quality Level

mol wt

average Mn 900

Mw/Mn

<1.1

Ω-end

tosylate

α-end

methoxy

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service