Skip to Content
Merck
All Photos(1)

Key Documents

729108

Sigma-Aldrich

Poly(ethylene glycol) methyl ether thiol

average MN 800, chemical modification reagent thiol reactive, methoxy, thiol

Synonym(s):

Polyethylene glycol, Methoxy PEG thiol, Methoxypolyethylene glycol thiol, PEG thiol, mPEG thiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)nCH2CH2SH
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

Poly(ethylene glycol) methyl ether thiol, average Mn 800

form

solid

mol wt

average Mn 800

reaction suitability

reagent type: chemical modification reagent
reactivity: thiol reactive

impurities

≤10% disulfide

transition temp

Tm 32-37 °C

Mw/Mn

≤1.1

Ω-end

thiol

α-end

methoxy

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Poly(ethylene glycol) methyl ether thiol can be used to functionalize Fe3O4@Au core-shell nanoparticles. The thiolated PEG increases the water solubility and significantly lowers the nanotoxicity.

It can be used as a precursor to synthesize PEG conjugated poly(β-amino ester) polyplexes for gene therapy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Gentilini, Mariangela Boccalon, and Lucia Pasquato
The Journal of Organic Chemistry, 3308-3308 (2008)
Nisha C. Kalarickal, Stephen Rimmer, Prodip Sarker, and Jean-Christophe Leroux
Macromolecules, 40, 1874-1874 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service