726184
trans-4-Cyclohexene-1,2-diamine dihydrochloride
≥98.0%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H12N2 · 2HCl
CAS Number:
Molecular Weight:
185.09
Beilstein:
3124971
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥98.0%
98.0-102.0% (AT)
form
powder
SMILES string
Cl.Cl.N[C@@H]1CC=CC[C@H]1N
InChI
1S/C6H12N2.2ClH/c7-5-3-1-2-4-6(5)8;;/h1-2,5-6H,3-4,7-8H2;2*1H/t5-,6-;;/m1../s1
InChI key
UQRRHSHMGMLJDM-BNTLRKBRSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
W.C.M.C. Kokke, F.A. Varkevisser
The Journal of Organic Chemistry, 39, 1535-1539 (1974)
A. Burgos, G.J. Ellames
Journal of Labelled Compounds & Radiopharmaceuticals, XLI, 443-449 (1998)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service