Skip to Content
Merck
All Photos(2)

Documents

725528

Sigma-Aldrich

Tetrakis(ethylmethylamido)zirconium(IV)

packaged for use in deposition systems

Synonym(s):

TEMAZ, Tetrakis(ethylmethylamino)zirconium(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr(NCH3C2H5)4
CAS Number:
Molecular Weight:
323.63
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

reaction suitability

core: zirconium
reagent type: catalyst

bp

81 °C/0.1 mmHg (lit.)

density

1.049 g/mL at 25 °C (lit.)

SMILES string

CCN(C)[Zr](N(C)CC)(N(C)CC)N(C)CC

InChI

1S/4C3H8N.Zr/c4*1-3-4-2;/h4*3H2,1-2H3;/q4*-1;+4

InChI key

SRLSISLWUNZOOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 40 Zirconium

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Won, S-J.; Kim, J-Y.; Choi, G-J.; Heo, J.; Hwang, C. S.; Kim, H.
Journal of Materials Chemistry, 21, 4374-4374 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service