72351
Benzyl nicotinate
≥98.0% (GC)
Synonym(s):
Nicotinic acid benzyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C13H11NO2
CAS Number:
Molecular Weight:
213.23
Beilstein:
159169
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥98.0% (GC)
form
liquid
refractive index
n20/D 1.570
bp
177 °C/8 mmHg (lit.)
mp
24 °C (lit.)
functional group
ester
phenyl
SMILES string
O=C(OCc1ccccc1)c2cccnc2
InChI
1S/C13H11NO2/c15-13(12-7-4-8-14-9-12)16-10-11-5-2-1-3-6-11/h1-9H,10H2
InChI key
KVYGGMBOZFWZBQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Benzyl nicotinate is a benzyl ester. It is a vasodilator agent and is also generally used in cosmetics and drugs.
Application
Benzyl nicotinate can be used for the synthesis of benzylpalladium complexes to be used as catalysts for the substitution of olefins with benzylic groups.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Zrinka Abramovic et al.
Advances in experimental medicine and biology, 701, 75-82 (2011-03-30)
Tumor hypoxia is a well known therapeutic problem which contributes to radioresistance and aggressive tumor characteristics. Lack of techniques for repeated measurements of tumor oxygenation (pO(2), partial pressure of oxygen) has restricted the optimization of hypoxia modifying methods and their
Synthesis of benzylpalladium complexes through C-O bond cleavage of benzylic carboxylates: Development of a novel palladium-catalyzed benzylation of olefins.
Narahashi H, et al.
Journal of Organometallic Chemistry, 693(2), 283-296 (2008)
V Erjavec et al.
International journal of pharmaceutics, 307(1), 1-8 (2005-11-01)
The purpose of this study was to select the best types of liposomes for use as drug carriers for topical treatment of oral mucosal lesions. Electron paramagnetic resonance (EPR) oximetry, using the paramagnetic probe lithium phthalocyanine, was used in vivo
Efficient transesterification/acylation reactions mediated by N-heterocyclic carbene catalysts.
Grasa GA, et al.
The Journal of Organic Chemistry, 68(7), 2812-2819 (2003)
Julijana Kristl et al.
AAPS pharmSci, 5(1), E2-E2 (2003-04-26)
New and improved drug delivery systems are the important subject of much scientific research. The development of formulations that increase skin oxygenation and of methods for measuring oxygen levels in skin are important for dealing with healing processes affected by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service