717592
2-Chloroadenine
96%
Synonym(s):
2-Chloro-6-aminopurine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H4ClN5
CAS Number:
Molecular Weight:
169.57
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
96%
form
solid
mp
384-389 °C
functional group
chloro
SMILES string
Nc1nc(Cl)nc2nc[nH]c12
InChI
1S/C5H4ClN5/c6-5-10-3(7)2-4(11-5)9-1-8-2/h1H,(H3,7,8,9,10,11)
InChI key
HBJGQJWNMZDFKL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
P Hentosh et al.
Nucleic acids research, 19(11), 3143-3148 (1991-06-11)
The purine analog, 2-chloro-2'-deoxyadenosine triphosphate (CldATP), was incorporated enzymatically in place of dATP into the minus strand of M13mp18 duplex DNA. Its effect on protein-DNA interactions was assessed by determining the amount of DNA cleavage by type II restriction endonucleases.
Peng Li et al.
Biochemical pharmacology, 79(7), 1015-1024 (2009-11-10)
Since the heterologous tolerance that develops after chronic morphine administration has been proposed to be an adaptive process, it follows that the time course of the change in the cellular components should coincide with the time course of the altered
Kinga Banach et al.
Nucleosides, nucleotides & nucleic acids, 24(5-7), 465-469 (2005-10-27)
A new kinetic model is presented for analysis of experimental data of oxidation process catalyzed by milk xanthine oxidase. The kinetics for two substrates, xanthine and its analog 2-chloroadenine, in a broad pH range (5.8-9.0) are best described by an
Laura Willems et al.
Journal of molecular and cellular cardiology, 38(2), 245-256 (2005-02-09)
Aging is associated with reduced tolerance to ischemic insult, and genesis of this intolerant phenotype is poorly understood. We characterized effects of aging and gender on cardiovascular function and cell damage during 20 min ischemia and 60 min reperfusion in
S A Taran et al.
Prikladnaia biokhimiia i mikrobiologiia, 44(2), 181-186 (2008-08-02)
Cladribine (2-chloro-2'-deoxyadenosine) was synthesized using intact cells of the recombinant Escherichia coli strain producing Geobacillus stearothermophilus B-2194 thermostable purine-nucleoside phosphorylase II (EC 2.4.2.1). Use of the cells containing this thermostable enzyme allowed the process to be conducted at a temperature
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service