Skip to Content
Merck
All Photos(1)

Key Documents

714917

Sigma-Aldrich

Ethylbenzene solution

NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10
CAS Number:
Molecular Weight:
106.17
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.12

grade

NMR reference standard

form

solution

composition

TMS, 0.01%

concentration

0.1% in chloroform-d (99.8 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in. , ultra-thin wall

NMR tube wall thickness

0.24 mm

suitability

suitable for NMR (reference standard)

SMILES string

CCc1ccccc1

InChI

1S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

YNQLUTRBYVCPMQ-UHFFFAOYSA-N

Related Categories

Features and Benefits

1H sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Rezazadeh Azari et al.
The international journal of occupational and environmental medicine, 3(1), 39-44 (2012-10-02)
Benzene, toluene, ethylbenzene and xylene (BTEX) are the most important toxic volatile compounds in the air and could be easily absorbed through the respiratory tract. In recent years, the risk of exposure to BTEX compounds, especially benzene as a carcinogen
You-Ya Zhou et al.
Environmental monitoring and assessment, 185(4), 3037-3048 (2012-09-11)
A method based on headspace (HS) sampling coupling with portable gas chromatography (GC) with photo ionization detector (PID) was developed for rapid determination of benzene, toluene, ethylbenzene, and xylenes (BTEX) in soils. Optimal conditions for HS gas sampling procedure were
Ch Vlachokostas et al.
Environmental pollution (Barking, Essex : 1987), 177, 125-134 (2013-03-19)
Emission from road traffic has become the most important source of local air pollution in numerous European cities. Epidemiological research community has established consistent associations between traffic-related substances and various health outcomes. Nevertheless, the vast majority of urban areas are
Carla B Vidal et al.
Journal of environmental management, 112, 178-185 (2012-08-28)
Various technologies have been used for the treatment and remediation of areas contaminated by BTEX (benzene, toluene, ethylbenzene and xylenes), which are organic compounds that are of particular concern due to their toxicity. Potential applications of synthetic zeolites for environmental
T Tunsaringkarn et al.
The international journal of occupational and environmental medicine, 3(3), 117-125 (2012-10-02)
Gasoline station workers are exposed to volatile organic compounds such as benzene, toluene, ethylbenzene and xylene (BTEX). To determine the level of exposure to BTEX compounds among gasoline station workers and measure the roadside concentrations of these compounds in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service