Skip to Content
Merck
All Photos(1)

Documents

713643

Sigma-Aldrich

N-Boc-cis-4-azido-L-proline (dicyclohexylammonium) salt

≥98.0%

Synonym(s):

(2S,4S)-N-Boc-4-azidopyrrolidine-2-carboxylic acid (dicyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H39N5O4
CAS Number:
Molecular Weight:
437.58
Beilstein:
5977215
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)
≥98.0%
98.0-102.0% (NT)

form

lumps

optical activity

[α]/D -10.0±1.0°, c = 1 in ethanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

C1CCC(CC1)NC2CCCCC2.CC(C)(C)OC(=O)N3C[C@H](C[C@H]3C(O)=O)N=[N+]=[N-]

InChI

1S/C12H23N.C10H16N4O4/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-10(2,3)18-9(17)14-5-6(12-13-11)4-7(14)8(15)16/h11-13H,1-10H2;6-7H,4-5H2,1-3H3,(H,15,16)/t;6-,7-/m.0/s1

InChI key

XYLCLQGQCINCLJ-BLOLRMMYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service