Skip to Content
Merck
All Photos(1)

Key Documents

704709

Sigma-Aldrich

p-Terphenyl-4,4′′-dithiol

95%

Synonym(s):

1,4-Bis(4-mercaptophenyl)benzene, TPDT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSC6H4C6H4C6H4SH
CAS Number:
Molecular Weight:
294.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

mp

294-299 °C

storage temp.

−20°C

SMILES string

Sc1ccc(cc1)-c2ccc(cc2)-c3ccc(S)cc3

InChI

1S/C18H14S2/c19-17-9-5-15(6-10-17)13-1-2-14(4-3-13)16-7-11-18(20)12-8-16/h1-12,19-20H

InChI key

HSAKNFPXLQYFIJ-UHFFFAOYSA-N

General description

p-Terphenyl-4,4″ -dithiol (TPDT) is an alkanethiol that forms a self-assembled monolayer (SAM) on a variety of substrates. It can be used to form a dithiol ligand that functionalizes and modifies the properties of surfaces.

Application

TPDT forms a SAM on gold nanoparticle by immobilizing the surface atoms, which can be used in opto-electronic and other energy based applications.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Raman Spectroscopy of Biphenyl-4, 4′-dithiol and p-Terphenyl-4, 4″ -dithiol on Gold Surfaces.
Ganbold E and Joo S
Bull. Korean Chem. Soc., 36(3), 887-890 (2015)
Photocatalytic activity and fluorescence of gold/zinc oxide nanoparticles formed by dithiol linking.
Unlu I, et al.
Langmuir, 31(31), 8718-8725 (2015)
Transition metal-free graphene framework based on disulfide bridges as a Li host material.
Yoo YG, et al.
Energy Storage Materials, 14(31), 238-245 (2018)
Roya Afsharipour et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 238, 118427-118427 (2020-05-11)
Molecularly imprinted polymer (MIP) was synthesized through the coprecipitation method on the graphene oxide anchored pencil lead as a substrate for the first time and applied as an efficient sorbent for pseudo stir bar sorptive extraction of nabumetone. The extracted
Simultaneous probing of nanocrystal (NC)-ligand interaction-induced charge transfer/transport properties at the electron donor (lead selenide NC)/acceptor (zinc oxide) functional interface.
Park B, et al.
Physica B: Condensed Matter, 553(31), 40-46 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service