Skip to Content
Merck
All Photos(1)

Key Documents

704296

Sigma-Aldrich

Lanthanum strontium manganite

greener alternative

LSM-20, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.23

Quality Level

Assay

≥99%

form

powder

mol wt

Mw 231.58 g/mol

composition

La0.8Sr0.2MnO3

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

battery manufacturing

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

LSM-20 is a state of art electrode material for solid oxide fuel cells (SOFCs). LSM-20 possesses both ionic and electronic conductivity and belongs to the ABO3 perovskite oxide family.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Lanthanum strontium manganite (LSM-20/GDC 10) is a ferromagnetic material, a Sr-doped LaMnO3 that is mainly used as a cathode material for solid oxide fuel cells (SOFCs). It also acts as a magnetic fluid, which can further be used as an inducer of tumor hyperthermia.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in Materials for Solid Oxide Fuel Cells
Gorte RJ
Material Matters, 5(4) null
Development of lanthanum strontium manganite perovskite cathode materials of solid oxide fuel cells: a review
Jiang SP
Chemosphere, 43, 6799-6833 (2008)
Aqueous slurry processing of monolithic films for SOFC-YSZ, LSM and YSZ-NiO systems
Ramanathan S and Kakade MB
International Journal of Hydrogen Energy, 36(22), 14956-14962 (2011)
Synthesis and characterization of lanthanum strontium manganite
Ghosh A, et al.
Scr. Mater., 52(12), 1305-1309 (2005)
Development of lanthanum strontium manganite perovskite cathode materials of solid oxide fuel cells: a review.
Jiang SP
J. Mater. Sci., 43(21), 6799-6833 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service