All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C6H3FN2
CAS Number:
Molecular Weight:
122.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
35-41 °C
functional group
fluoro
nitrile
SMILES string
Fc1ccc(nc1)C#N
InChI
1S/C6H3FN2/c7-5-1-2-6(3-8)9-4-5/h1-2,4H
InChI key
BHXHRMVSUUPOLX-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
210.0 °F
Flash Point(C)
98.9 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service