Skip to Content
Merck
All Photos(2)

Key Documents

703540

Sigma-Aldrich

2,2,6,6-Tetramethylpiperidinylmagnesium chloride lithium chloride complex solution

1.0 M in THF/toluene

Synonym(s):

TMPMgCl·LiCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18Cl2LiMgN
CAS Number:
Molecular Weight:
242.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in THF/toluene
20 % (w/w)

density

0.96 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li+].[Cl-].CC1(C)CCCC(C)(C)N1[Mg]Cl

InChI

1S/C9H18N.2ClH.Li.Mg/c1-8(2)6-5-7-9(3,4)10-8;;;;/h5-7H2,1-4H3;2*1H;;/q-1;;;+1;+2/p-2

InChI key

JHBZAAACZVPPRQ-UHFFFAOYSA-L

Application

2,2,6,6-Tetramethylpiperidinylmagnesium chloride lithium chloride complex is a non-nucleophilic Knochel-Hauser base mainly used for the magnesiation of functionalized arenes and heteroarenes to prepare the corresponding Grignard reagents. It can also be used as a base in the synthesis of Π-conjugated polymers via catalyst-transfer polycondensation.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Product of Albemarle US Inc
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

5.0 °F

Flash Point(C)

-15 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct, regioselective N-alkylation of 1, 3-azoles.
Chen S
Organic Letters, 18(1), 16-19 (2015)
Highly Functionalized Benzene Syntheses by Directed Mono or Multiple Magnesiations with TMPMgCl.LiCl.
Lin W
Organic Letters, 8(24), 5673-5676 (2006)
Scaleable preparation of functionalized organometallics via directed ortho metalation using Mg-and Zn-amide bases.
Wunderlich SH
Organic Process Research & Development, 14(2), 339-345 (2010)
Hiroki Iguchi et al.
Scientific reports, 7, 39937-39937 (2017-01-07)
Processing and manipulation of highly conductive pristine graphene in large quantities are still major challenges in the practical application of graphene for electric device. In the present study, we report the liquid-phase exfoliation of graphite in toluene using well-defined poly(3-hexylthiophene)
Conjugated Polymers with Repeated Sequences of Group 16 Heterocycles Synthesized through Catalyst-Transfer Polycondensation.
Tsai CH
Journal of the American Chemical Society, 138(21), 6798-6804 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service