Skip to Content
Merck
All Photos(1)

Key Documents

703206

Sigma-Aldrich

cis-Bis(isothiocyanato)bis(2,2′-bipyridyl-4,4′-dicarboxylato)ruthenium(II)

Synonym(s):

Greatcell Solar®, N-3 dye

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H16N6O8RuS2
CAS Number:
Molecular Weight:
705.64
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95% (NMR)

Quality Level

form

powder

mp

>300 °C

λmax

534, 395, 312 nm (lit.)

SMILES string

S=C=N[Ru]N=C=S.OC(=O)c1ccnc(c1)-c2cc(ccn2)C(O)=O.OC(=O)c3ccnc(c3)-c4cc(ccn4)C(O)=O

InChI

1S/2C12H8N2O4.2CNS.Ru/c2*15-11(16)7-1-3-13-9(5-7)10-6-8(12(17)18)2-4-14-10;2*2-1-3;/h2*1-6H,(H,15,16)(H,17,18);;;/q;;2*-1;+2

InChI key

VMISXESAJBVFNH-UHFFFAOYSA-N

General description

cis-Bis(isothiocyanato)bis(2,2′-bipyridyl-4,4′-dicarboxylato)ruthenium(II) (N3 dye) is a ruthenium polypyridyl based complex that can be used as a sensitizer. It is a highly stable polymer in which the photocurrent decreases with an increase in the concentration of N3 dye.

Application

N3 dye can be coated on electrodes for improving the sensitivity of the dye sensitized solar cells (DSSCs) which can further enhance the power conversion efficiency (PCE).

Legal Information

Product of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Dyesol is a registered trademark of Greatcell Solar

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stark effects after excited-state interfacial electron transfer at sensitized TiO2 nanocrystallites
Ardo S, et al.
Journal of the American Chemical Society, 132(19), 6696-6709 (2010)
Ruthenium (II) Charge-Transfer Sensitizers Containing 4, 4 `-Dicarboxy-2, 2 `-bipyridine. Synthesis, Properties, and Bonding Mode of Coordinated Thio-and Selenocyanates
Kohle O, et al.
Inorganic Chemistry, 35(16), 4779-4787 (1996)
Determination of the electron lifetime in nanocrystalline dye solar cells by open-circuit voltage decay measurements
Zaban A, et al.
ChemPhysChem, 4(8), 859-864 (2003)
Influence of electrolyte in transport and recombination in dye-sensitized solar cells studied by impedance spectroscopy
Fabregat-Santiago F, et al.
Solar Energy Materials and Solar Cells, 87(1-4), 117-131 (2005)
Transport-limited recombination of photocarriers in dye-sensitized nanocrystalline TiO2 solar cells
Kopidakis N, et al.
The Journal of Physical Chemistry B, 107(41), 11307-11315 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service