Skip to Content
Merck
All Photos(1)

Key Documents

696463

Sigma-Aldrich

5-Methoxy-6-azaindole

96%

Synonym(s):

5-Methoxy-1H-pyrrolo[2,3-c]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2O
CAS Number:
Molecular Weight:
148.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

powder

mp

122-127 °C

storage temp.

2-8°C

SMILES string

COc1cc2cc[nH]c2cn1

InChI

1S/C8H8N2O/c1-11-8-4-6-2-3-9-7(6)5-10-8/h2-5,9H,1H3

InChI key

CCNJNELOBGXDFD-UHFFFAOYSA-N

Related Categories

Application

Reactant for preparation of indole sulfonamides as HIV entry inhibitors
Reactant for synthesis of melatoninergic ligands including azaindole moiety

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service