693774
Chiralyst P1271
Umicore
Synonym(s):
Bis(1,5-cyclooctadiene)iridium(I) tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, Bis(cyclooctadiene)iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenyl)borate
About This Item
Recommended Products
form
solid
reaction suitability
core: iridium
reagent type: catalyst
SMILES string
[Ir+].C1CC=CCCC=C1.C2CC=CCCC=C2.FC(F)(F)c3cc(cc(c3)C(F)(F)F)[B-](c4cc(cc(c4)C(F)(F)F)C(F)(F)F)(c5cc(cc(c5)C(F)(F)F)C(F)(F)F)c6cc(cc(c6)C(F)(F)F)C(F)(F)F
InChI
1S/C32H12BF24.2C8H12.Ir/c34-25(35,36)13-1-14(26(37,38)39)6-21(5-13)33(22-7-15(27(40,41)42)2-16(8-22)28(43,44)45,23-9-17(29(46,47)48)3-18(10-23)30(49,50)51)24-11-19(31(52,53)54)4-20(12-24)32(55,56)57;2*1-2-4-6-8-7-5-3-1;/h1-12H;2*1-2,7-8H,3-6H2;/q-1;;;+1/b;2*2-1-,8-7-;
InChI key
YQXKTVMJDKWGHI-AUUWQFPRSA-N
Application
- Asymmetric hydrogenation reactions
- Directed cyclodehydration reaction
- Direct prenylation via C-H oxidative addition-allene insertion
- Double intramolecular hydroalkoxylation
- Intramolecular hydroamination reactions
- Directed C-H bond cleavage
- Imine vinylation reactions
Umicore Precatalysts for Asymmetric and Cross-Coupling Catalysis
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service