Skip to Content
Merck
All Photos(1)

Key Documents

692204

Sigma-Aldrich

(R)-[(RuCl(SEGPHOS®))2(μ-Cl)3][NH2Me2]

Synonym(s):

Dimethylammonium dichlorotri(μ-chloro)bis[(R)-(+)-5,5′-bis(diphenylphosphino)-4,4′-bi-1,3-benzodioxole]diruthenate(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH2+[C76H56Cl5O8P4Ru2]-
CAS Number:
Molecular Weight:
1646.64
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

functional group

amine
phosphine

storage temp.

2-8°C

SMILES string

Cl.[Cl-].CNC.[Cl+]1[Ru-]2[Cl+][Ru-]1[Cl+]2.C3Oc4ccc(P(c5ccccc5)c6ccccc6)c(c4O3)-c7c8OCOc8ccc7P(c9ccccc9)c%10ccccc%10.C%11Oc%12ccc(P(c%13ccccc%13)c%14ccccc%14)c(c%12O%11)-c%15c%16OCOc%16ccc%15P(c%17ccccc%17)c%18ccccc%18

InChI

1S/2C38H28O4P2.C2H7N.2ClH.3Cl.2Ru/c2*1-5-13-27(14-6-1)43(28-15-7-2-8-16-28)33-23-21-31-37(41-25-39-31)35(33)36-34(24-22-32-38(36)42-26-40-32)44(29-17-9-3-10-18-29)30-19-11-4-12-20-30;1-3-2;;;;;;;/h2*1-24H,25-26H2;3H,1-2H3;2*1H;;;;;/q;;;;;3*+1;2*-1/p-1

InChI key

RENFEASRVBXAEN-UHFFFAOYSA-M

General description

(R)-[(RuCl(SEGPHOS®))2(μ-Cl)3][NH2Me2] is a ruthenium-based catalyst generally used in the asymmetric hydrogenation reactions of ketones. SEGPHOS®, the chiral ligand and its ruthenium-based catalysts are developed and commercialized by Takasago International Corporation.

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 3148136, 3549390
SEGPHOS is a registered trademark of Takasago Intl. Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Developments in asymmetric hydrogenation from an industrial perspective
Shimizu H, et al.
Accounts of Chemical Research, 40(12), 1385-1393 (2007)
Asymmetric catalysis by architectural and functional molecular engineering: practical chemo-and stereoselective hydrogenation of ketones
Noyori R and Ohkuma T
Angewandte Chemie (International ed. in English), 40(1), 40-73 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service