Skip to Content
Merck
All Photos(1)

Key Documents

688606

Sigma-Aldrich

trans-2-Methyl-2-butenoyl chloride

≥97.0%

Synonym(s):

(E)-2,3-Dimethylacryloyl chloride, (E)-2-Methylcrotonoyl chloride, 3-cis-Methyl-methacryloyl chloride, Tigloyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7ClO
CAS Number:
Molecular Weight:
118.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0%

form

liquid

refractive index

n20/D 1.471

functional group

acyl chloride

storage temp.

−20°C

SMILES string

C\C=C(/C)C(Cl)=O

InChI

1S/C5H7ClO/c1-3-4(2)5(6)7/h3H,1-2H3/b4-3+

InChI key

TUNLYEHIVPWOHK-ONEGZZNKSA-N

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service