682659
Nanoclay, hydrophilic bentonite
Synonym(s):
Nanomer® PGV, Nanomer® clay, Montmorillinite clay, bentonite
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
powder
Quality Level
particle size
≤25 μm
SMILES string
[Si](=O)=O.[Si](=O)=O.[Si](=O)=O.[Si](=O)=O.[Al]21O[Al](O2)O1.O
InChI key
KARVSHNNUWMXFO-UHFFFAOYSA-N
Application
For representative application data and nanocomposite processing methods see datasheet G105.
Untreated (no organic modification) hydrophilic clay for dispersion in water-based polymers and coatings
- Rheology control
- Increase barrier
- Enhance chemical resistance
Legal Information
Nanomer® clays are products of Nanocor Corporation
Product of Nanocor®, Inc.
Product of Nanocor®, Inc.
Nanomer is a registered trademark of Amcol International Corporation
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Zoleikha Hajizadeh et al.
Scientific reports, 10(1), 11671-11671 (2020-07-17)
Geopolymers as aluminosilicate inorganic polymers and eco-friendly building materials which can be used as substrate for different kinds of composite. In this research, according to the fabrication of geopolymer based on bentonite as a substrate and embedment of NiFe2O4 nanoparticles
C H Hu et al.
Poultry science, 92(1), 143-150 (2012-12-18)
A total of 450 one-day-old Arbor Acres male chickens were used to investigate the effects of zinc oxide-montmorillonite hybrid (ZnO-MMT) on growth performance, intestinal structure, and function. The birds were allotted to 5 dietary treatments for 21 d, each of
A Christy Wyckoff et al.
PloS one, 8(3), e58630-e58630 (2013-03-14)
Prions, the infectious agent of scrapie, chronic wasting disease and other transmissible spongiform encephalopathies, are misfolded proteins that are highly stable and resistant to degradation. Prions are known to associate with clay and other soil components, enhancing their persistence and
Xue Jin et al.
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences, 41(6), 631-638 (2012-12-15)
To develop polyethylenimine-Doxorubicin-montmorillonite (PEI-Dox-MTT) as a novel multifunction delivery system. Dox was intercalated into montmorillonite, PEI covered to the surface of Dox/MMT to make the nano-particle. XRD, FT-IR and TGA were used to confirm chemical property of the nano-particle. SEM
Katarzyna Majchrzycka et al.
International journal of occupational safety and ergonomics : JOSE, 18(3), 375-385 (2012-09-22)
This study evaluated the bioactivity of polypropylene melt-blown filtering nonwovens used in respiratory protective devices (RPD) with a biocidal agent (alkylammonium microbiocides) on 2 mineral carriers. Two types of carriers were tested: a bentonite, with an aluminosilicate base, and a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service