682101
Bis(trimethylaluminum)-1,4-diazabicyclo[2.2.2]octane adduct
Synonym(s):
Bis(trimethylaluminum)-DABCO® adduct, DABAL-Me3, DABAL-trimethylaluminum, Triethylenediaminine bis(trimethylaluminum)
About This Item
Recommended Products
mp
>250 °C
Quality Level
SMILES string
C[Al-](C)(C)[N+]12CC[N+](CC1)(CC2)[Al-](C)(C)C
InChI
1S/C6H12N2.6CH3.2Al/c1-2-8-5-3-7(1)4-6-8;;;;;;;;/h1-6H2;6*1H3;;
InChI key
WSTAITCRSVOCTK-UHFFFAOYSA-N
Related Categories
General description
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 1
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service