Skip to Content
Merck
All Photos(2)

Key Documents

679305

Sigma-Aldrich

Triethoxy(octyl)silane

deposition grade, 97%, 99.99% trace metals basis

Synonym(s):

Octyltriethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7Si(OC2H5)3
CAS Number:
Molecular Weight:
276.49
Beilstein:
2325287
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

Quality Level

Assay

97%
99.99% trace metals basis

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

84-85 °C/0.5 mmHg (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC[Si](OCC)(OCC)OCC

InChI

1S/C14H32O3Si/c1-5-9-10-11-12-13-14-18(15-6-2,16-7-3)17-8-4/h5-14H2,1-4H3

InChI key

MSRJTTSHWYDFIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Sun et al.
Talanta, 143, 127-131 (2015-06-17)
The sensitivity of pH indicators is always confined in acidic solvents used during the immobilization because of the large deviation from their color transition points. By regulating the pH of indicators using organic acid or base, the color transition points
Yueer Yan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(34), 12161-12170 (2015-07-16)
A clear and deep understanding of zeolite crystallization with the addition of organosilane is desirable for the reasonable design and preparation of hierarchical zeolites. Herein, the effects of different organosilanes on zeolite crystallization were systematically studied. It was found that
Gulsah Ozan Aydin et al.
Marine pollution bulletin, 96(1-2), 155-164 (2015-05-24)
In this study, reusable poly(alkoxysilane) organogels with high absorption capacities were synthesized by the condensation of a cyclo aliphatic glycol (UNOXOL™) and altering the chain length of the alkyltriethoxysilanes. The structural and thermal properties of cross-linked poly(alkoxysilane) polymers were determined
Andrea Penna et al.
Journal of the American Society for Mass Spectrometry, 26(8), 1311-1319 (2015-04-22)
Since it was proposed for the first time, desorption electrospray ionization-mass spectrometry (DESI-MS) has been evaluated for applicability in numerous areas. Elucidations of the ionization mechanisms and the subsequent formation of isolated gas-phase ions have been proposed so far. In
Deposition of wafer-scale single-component and binary nanocrystal superlattice thin films via dip-coating.
E Ashley Gaulding et al.
Advanced materials (Deerfield Beach, Fla.), 27(18), 2846-2851 (2015-03-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service