Skip to Content
Merck
All Photos(2)

Key Documents

675938

Sigma-Aldrich

Me4tButylXphos

96%

Synonym(s):

2-Di-tert-butylphosphino-3,4,5,6-tetramethyl-2′,4′,6′-triisopropyl-1,1′-biphenyl, Tetramethyl di-tBuXPhos, Tetramethyl tBuXPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H53P
CAS Number:
Molecular Weight:
480.75
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

96%

form

solid

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: C-X Bond Formation

mp

168-172 °C

functional group

phosphine

SMILES string

CC1=C(C)C(C)=C(C)C(C(C(C(C)C)=CC(C(C)C)=C2)=C2C(C)C)=C1P(C(C)(C)C)C(C)(C)C

InChI

1S/C33H53P/c1-19(2)26-17-27(20(3)4)30(28(18-26)21(5)6)29-24(9)22(7)23(8)25(10)31(29)34(32(11,12)13)33(14,15)16/h17-21H,1-16H3

InChI key

RCRYEYMHBHPZQD-UHFFFAOYSA-N

Legal Information

Usage subject to US Patents 6307087 and 6395916.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Ikawa et al.
Journal of the American Chemical Society, 129(43), 13001-13007 (2007-10-09)
We present results on the amidation of aryl halides and sulfonates utilizing a monodentate biaryl phosphine-Pd catalyst. Our results are in accord with a previous report that suggests that the formation of kappa(2)-amidate complexes is deleterious to the effectiveness of
Huy Tran et al.
Angewandte Chemie (International ed. in English), 61(1), e202110575-e202110575 (2021-10-30)
A one-pot cascade sequence to generate synthetically challenging polycyclic scaffolds is reported utilizing a novel Lewis acid gold catalyst for the key cyclization step, enabling the divergent synthesis of both 6,6,5-tricyclic and 6,6,6,5-tetracyclic cores through both ligand and reaction condition
N Ian Rinehart et al.
Science (New York, N.Y.), 381(6661), 965-972 (2023-08-31)
Machine-learning methods have great potential to accelerate the identification of reaction conditions for chemical transformations. A tool that gives substrate-adaptive conditions for palladium (Pd)-catalyzed carbon-nitrogen (C-N) couplings is presented. The design and construction of this tool required the generation of
Kevin W Anderson et al.
Journal of the American Chemical Society, 128(33), 10694-10695 (2006-08-17)
The direct and selective synthesis of phenols from aryl/heteroaryl halides and KOH has been achieved through the use of highly active monophosphine-based catalysts derived from Pd(2)dba(3) and ligands L1 or L2 and the biphasic solvent system 1,4-dioxane/H(2)O. We have also
David S Surry et al.
Angewandte Chemie (International ed. in English), 47(34), 6338-6361 (2008-07-30)
Palladium-catalyzed amination reactions of aryl halides have undergone rapid development in the last 12 years, largely driven by the implementation of new classes of ligands. Biaryl phosphanes have proven to provide especially active catalysts in this context. This Review discusses

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service