Skip to Content
Merck
All Photos(1)

Key Documents

675105

Sigma-Aldrich

(11-Mercaptoundecyl)hexa(ethylene glycol)

90%

Synonym(s):

(1-Mercaptoundec-11-yl)hexa(ethylene glycol), Hexa(ethylene glycol)mono-11-mercaptoundecyl ether, MUHEG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H48O7S
CAS Number:
Molecular Weight:
468.69
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

90%

refractive index

n20/D 1.474

density

1.0154 g/mL at 25 °C

storage temp.

−20°C

SMILES string

OCCOCCOCCOCCOCCOCCOCCCCCCCCCCCS

InChI

1S/C23H48O7S/c24-10-12-26-14-16-28-18-20-30-22-21-29-19-17-27-15-13-25-11-8-6-4-2-1-3-5-7-9-23-31/h24,31H,1-23H2

InChI key

QYKSUHRPPSCIFK-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service