Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
674397
11-Amino-1-undecanethiol hydrochloride
97%
Synonym(s):
AUT, 11-Aminoundecanethiol hydrochloride, 11-Mercaptoundecylamine, MUAM
About This Item
Recommended Products
Quality Level
Assay
97%
form
solid
mp
120-170 °C
storage temp.
2-8°C
SMILES string
Cl[H].NCCCCCCCCCCCS
InChI
1S/C11H25NS.ClH/c12-10-8-6-4-2-1-3-5-7-9-11-13;/h13H,1-12H2;1H
InChI key
LOQACAFAHHWTBJ-UHFFFAOYSA-N
General description
Application
It can also be self-assembled onto gold electrodes for a study on the effect of terminal groups on the redox responses of ferrocene derivatives using the voltammetric method.[5]
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
-
Hi, I need to know the chemical stability of this item when in solution (i.e. ethanol). Due to its price we cannot discard the excess amounts made for each experiment. Is there any information on that please?
1 answer-
Unfortunately, the stability of this product in solution is not determined. Please see the link below to following article which may be helpful:
https://pubmed.ncbi.nlm.nih.gov/?term=11-amino-1-undecanethiol+hydrochloride+solutionHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service