Skip to Content
Merck
All Photos(1)

Key Documents

67073

Sigma-Aldrich

7-Methylguanine

≥98.0% (HPLC)

Synonym(s):

2-Amino-6-hydroxy-7-methylpurine, Epiguanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N5O
CAS Number:
Molecular Weight:
165.15
Beilstein:
174245
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

impurities

≤1% guanine

SMILES string

Cn1cnc2N=C(N)NC(=O)c12

InChI

1S/C6H7N5O/c1-11-2-8-4-3(11)5(12)10-6(7)9-4/h2H,1H3,(H3,7,9,10,12)

InChI key

FZWGECJQACGGTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chie Tomikawa et al.
Nucleic acids research, 38(3), 942-957 (2009-11-26)
N(7)-methylguanine at position 46 (m(7)G46) in tRNA is produced by tRNA (m(7)G46) methyltransferase (TrmB). To clarify the role of this modification, we made a trmB gene disruptant (DeltatrmB) of Thermus thermophilus, an extreme thermophilic eubacterium. The absence of TrmB activity
H A Billson et al.
Carcinogenesis, 30(4), 615-620 (2009-01-27)
Components of human diets may influence the incidence of colorectal adenomas, by modifying exposure or susceptibility to DNA-damaging alkylating agents. To examine this hypothesis, a food frequency questionnaire was used to assess the diet of patients recruited for a case-referent
Chiung-Wen Hu et al.
Chemical research in toxicology, 25(11), 2386-2392 (2012-10-05)
Areca nut is a carcinogen to humans and has been strongly associated with oral premalignant and malignant diseases. Previous studies speculated the presence of unknown direct-acting mutagens present in aqueous extracts of areca nut. We hypothesized whether any direct-acting alkylating
Kazuyoshi Tamae et al.
Cancer science, 100(4), 715-721 (2009-05-27)
Urinary 8-hydroxydeoxyguanosine (8-OH-dG) and 7-methylguanine (m7Gua) were measured by a column-switching high performance liquid chromatography method as markers of oxidative and methylating DNA damage, respectively. We investigated the associations between urinary 8-OH-dG or m7Gua and various lifestyle and demographic factors
Deepali Gupta et al.
Dalton transactions (Cambridge, England : 2003), (1)(1), 73-84 (2009-12-22)
In solution, unsubstituted guanine (GH2) is in a tautomeric equilibrium between 1H,7H-keto and 1H,9H-keto forms. Upon coordination with (dien)Pt(II), N9-bound mononuclear complex is preferentially formed. With excess (dien)Pt(II), the dinuclear complex [{Pt(dien)}2(GH-N7,N9)]3+ is formed at basic pH. The X-ray crystal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service