668435
1,2-Bis[(2S,5S)-2,5-diisopropylphospholano]ethane
kanata purity
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C22H44P2
CAS Number:
Molecular Weight:
370.53
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
form
solid
mp
67-72 °C
functional group
phosphine
SMILES string
CC(C)[C@H]1P(CCP2[C@H](C(C)C)CC[C@H]2C(C)C)[C@H](C(C)C)CC1
InChI
1S/C22H44P2/c1-15(2)19-9-10-20(16(3)4)23(19)13-14-24-21(17(5)6)11-12-22(24)18(7)8/h15-22H,9-14H2,1-8H3/t19-,20-,21-,22-/m1/s1
InChI key
IRVIFEWWPYKALC-GXRSIYKFSA-N
General description
1,2-Bis[(2S,5S)-2,5-diisopropylphospholano]ethane is a chiral C2-symmetric ligand, which on ligation with metal complexes forms active catalysts for asymmetric hydrogenation.
Legal Information
Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A mass spectrometric method for rapidly assaying the chiral selectivities of the copper (I) complexes of C2-symmetric ligands.
Davis MC and Gronert S.
Journal of Mass Spectrometry : Jms, 50(11), 1279-1287 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service