Skip to Content
Merck
All Photos(1)

Key Documents

668400

Sigma-Aldrich

1,3-Dimethylimidazolium-2-carboxylate

technical grade, ≥80%

Synonym(s):

1,3-Bis(methyl)imidazolium-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2
CAS Number:
Molecular Weight:
140.14
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

≥80%

form

solid

reaction suitability

reagent type: catalyst

mp

221-225 °C

storage temp.

2-8°C

SMILES string

Cn1cc[n+](C)c1C([O-])=O

InChI

1S/C6H8N2O2/c1-7-3-4-8(2)5(7)6(9)10/h3-4H,1-2H3

InChI key

HESZXGSSISDCNI-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Azouri M. et al.
European Journal of Inorganic Chemistry, 4877-4877 (2007)
Holbrey, J. D. et al.
Chemical Communications (Cambridge, England), 28-28 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service