Skip to Content
Merck
All Photos(1)

Documents

661074

Sigma-Aldrich

N-Boc-serinol

97%

Synonym(s):

1,1-Dimethylethyl [2-hydroxy-1-(hydroxymethyl)ethyl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17NO4
CAS Number:
Molecular Weight:
191.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

85-89 °C

SMILES string

CC(C)(C)OC(=O)NC(CO)CO

InChI

1S/C8H17NO4/c1-8(2,3)13-7(12)9-6(4-10)5-11/h6,10-11H,4-5H2,1-3H3,(H,9,12)

InChI key

JHBKBRLRYPYBLP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ritesh M Pabari et al.
International journal of pharmaceutics, 565, 533-542 (2019-05-16)
In this study we examined the potential of novel biodegradable polymers of polyesterurethane (PU), and its PEGylated (PU-PEG) form as nanocarriers of Infliximab (INF), to treat inflammation in an in-vitro epithelial model. Nanoparticles (NPs) formulated were of average size of
Krishna Madhavan et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 106(1), 278-290 (2017-01-29)
The optimization of biomechanical and biochemical properties of a vascular graft to render properties relevant to physiological environments is a major challenge today. These critical properties of a vascular graft not only regulate its stability and integrity, but also control
Alessandro Torchio et al.
Journal of materials chemistry. B, 8(34), 7696-7712 (2020-07-30)
The design of supramolecular (SM) hydrogels based on host-guest complexes represents an effective strategy to develop drug delivery systems. In this work, we designed SM hydrogels based on α-cyclodextrin and high-molar mass amphiphilic poly(ether urethane)s (PEUs, ) based on Poloxamer®
Monica Boffito et al.
Frontiers in bioengineering and biotechnology, 8, 384-384 (2020-06-09)
Injectable therapeutic formulations locally releasing their cargo with tunable kinetics in response to external biochemical/physical cues are gaining interest in the scientific community, with the aim to overcome the cons of traditional administration routes. In this work, we proposed an
Sharon Wei Ling Lee et al.
Advanced healthcare materials, 9(7), e1901486-e1901486 (2020-03-04)
Polymer nanoparticles (NPs), due to their small size and surface functionalization potential have demonstrated effective drug transport across the blood-brain-barrier (BBB). Currently, the lack of in vitro BBB models that closely recapitulate complex human brain microenvironments contributes to high failure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service