Skip to Content
Merck
All Photos(1)

Documents

656364

Sigma-Aldrich

(S)-(+)-3-Piperidinecarboxylic acid

97%

Synonym(s):

(S)-(+)-Nipecotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]/D 3.0 to 6.5°, c = 1 in H2O

mp

254 °C (dec.)

SMILES string

OC(=O)[C@H]1CCCNC1

InChI

1S/C6H11NO2/c8-6(9)5-2-1-3-7-4-5/h5,7H,1-4H2,(H,8,9)/t5-/m0/s1

InChI key

XJLSEXAGTJCILF-YFKPBYRVSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(-)-3-Piperidinecarboxylic acid is an inhibitor of GABA (γ-aminobutyric acid) uptake.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna-Lucia Koerling et al.
Neuropharmacology, 148, 394-405 (2018-11-26)
In addition to reducing seizures, anti-epileptic treatments should preserve physiological network activity. Here, we used a thalamocortical slice preparation displaying physiological slow oscillations to investigate the effects of anticonvulsant drugs on physiological activity and epileptiform activity in two pharmacological epilepsy
Inhibition of the uptake of GABA and related amino acids in rat brain slices by the optical isomers of nipecotic acid.
G A Johnston et al.
Journal of neurochemistry, 26(5), 1029-1032 (1976-05-01)
Carmela Napolitano et al.
Molecules (Basel, Switzerland), 14(9), 3268-3274 (2009-09-29)
As a novel example of improved entry of poorly delivered drugs into the brain by means of nutrient conjugates, L-carnitine was conjugated to nipecotic acid and the capacity to antagonize PTZ-induced convulsions of this novel entity was evaluated.
Moslem Mohammadi et al.
Toxicology, 244(1), 42-48 (2007-12-07)
A synaptosomal model was used to evaluate in vivo effects of paraoxon on the uptake of [(3)H]GABA in rat cerebral cortex and hippocampus. Male Wistar rats were given a single intraperitoneal injection of one of three doses of paraoxon (0.1
Robert A Darnall et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 302(5), R551-R560 (2011-12-14)
Arousal is an important defense against hypoxia during sleep. Rat pups exhibit progressive arousal impairment (habituation) with multiple hypoxia exposures. The mechanisms are unknown. The medullary raphe (MR) is involved in autonomic functions, including sleep, and receives abundant GABAergic inputs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service