Skip to Content
Merck
All Photos(1)

Key Documents

642622

Sigma-Aldrich

3-Pyridineboronic acid neopentylglycol ester

97%

Synonym(s):

5,5-Dimethyl-2-(3-pyridyl)-1,3,2-dioxaborinane, Pyridine-3-boronic acid neopentylglycol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14BNO2
CAS Number:
Molecular Weight:
191.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

92-96 °C (lit.)

SMILES string

CC1(C)COB(OC1)c2cccnc2

InChI

1S/C10H14BNO2/c1-10(2)7-13-11(14-8-10)9-4-3-5-12-6-9/h3-6H,7-8H2,1-2H3

InChI key

QMEKTOQBDDVVBE-UHFFFAOYSA-N

Application

3-Pyridineboronic acid neopentylglycol ester can be used as a reactant:
  • To synthesize N-alkyl-3-boronopyridinium salts by reacting with corresponding alkyl halides.
  • In the ruthenium-catalyzed diastereoselective C-H bond α-arylation of cyclic amines.
  • To prepare 2-(3-pyridyl)benzoxazole by reacting with benzoxazole via copper-catalyzed oxidative arylation reaction.
  • In the Suzuki-Miyaura cross-coupling reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-Catalyzed Oxidative Arylation of Heteroarenes under Mild Conditions Using Dioxygen as the Sole Oxidant
Yang F, et al.
Chemistry?A European Journal , 17(23), 6321-6325 (2011)
Nickel/N-Heterocyclic carbene-catalyzed Suzuki-Miyaura type cross-coupling of aryl carbamates
Ohtsuki A, et al.
The Journal of Organic Chemistry, 81(19), 9409-9414 (2016)
sp3 C- H bond arylation directed by amidine protecting group: α-arylation of pyrrolidines and piperidines
Pastine SJ, et al.
Journal of the American Chemical Society, 128(44), 14220-14221 (2006)
Preparation of a series of N-alkyl-3-boronopyridinium halides and study of their stability in the presence of hydrogen peroxide
Karpichev Y, et al.
Central European Journal of Chemistry, 10(4), 1059-1065 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service