Skip to Content
Merck
All Photos(1)

Documents

640573

Sigma-Aldrich

Cyclopropanesulfonyl chloride

95%

Synonym(s):

Cyclopropylsulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5ClO2S
CAS Number:
Molecular Weight:
140.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.4770 (lit.)

density

1.38 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClS(=O)(=O)C1CC1

InChI

1S/C3H5ClO2S/c4-7(5,6)3-1-2-3/h3H,1-2H2

InChI key

PFWWSGFPICCWGU-UHFFFAOYSA-N

Application

Cyclopropanesulfonyl chloride can be used in the synthesis of (1R,2S)-1-amino-2-vinylcyclopropanecarboxylic sulfonamide derivatives, which can serve as building blocks in the preparation of potent hepatitis C virus NS3 protease inhibitors.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Concise asymmetric synthesis of a (1R, 2S)-1-amino-2-vinylcyclopropanecarboxylic acid-derived sulfonamide and ethyl ester"
Lou Sha, et al.
Organic & Biomolecular Chemistry, 11(39), 6796-6805 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service