Skip to Content
Merck
All Photos(1)

Key Documents

631868

Supelco

1,4-Dioxane solution

NMR reference standard, 5 mM in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

grade

NMR reference standard
analytical standard

Quality Level

concentration

5 mM in chloroform-d (99.8 atom % D)

technique(s)

NMR: suitable

NMR tube size

3 mm × 8 in.

density

1.500 g/mL at 25 °C

format

single component solution

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

3 mm O.D. tube contains 0.235 mL.
Each pack contains 10 tubes.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos E Agudelo-Morales et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 13(18), 4195-4201 (2012-10-24)
The quenching of pyrene and 1-methylpyrene fluorescence by nitroanilines (NAs), such as 2-, 3-, and 4-nitroaniline (2-NA, 3-NA, and 4-NA, respectively), 4-methyl-3-nitroaniline (4-M-3-NA), 2-methyl-4-nitroaniline (2-M-4-NA), and 4-methyl-3,5-dinitroaniline (4-M-3,5-DNA), are studied in toluene and 1,4-dioxane. Steady-state fluorescence data show the higher
Iñaki Galve et al.
Molecular diversity, 16(4), 639-649 (2012-10-12)
A practical protocol was developed for the synthesis of 2-arylamino substituted 4-amino-5,6-dihydropyrido[2,3-d]pyrimidin-7(8H)-ones from α,β-unsaturated esters, malononitrile, and an aryl substituted guanidine via the corresponding 3-aryl-3,4,5,6- tetrahydropyrido[2,3-d]pyrimidin-7(8H)-ones. Such compounds are formed upon treatment of 2-methoxy-6-oxo-1,4,5,6-tetrahydropyridine-3-carbonitriles with an aryl substituted guanidine in
Kiyoaki Ishimoto et al.
Biomacromolecules, 13(11), 3757-3768 (2012-09-19)
For developing broader application of biobased polymers, graft copolymers and comb polymers having poly(lactic acid) (PLA) side chains have been synthesized by using a macromonomer technique. PLA macromonomers (MMm) having a methacryloyl polymerizable group with different PLA chain length with
R Dario Arrua et al.
Journal of chromatography. A, 1273, 26-33 (2013-01-01)
Rigid monolithic cryostructures were prepared in capillary format at sub-zero temperatures and used successfully in the separation of proteins by hydrophobic interaction chromatography (HIC). The polymerization mixture consisted of poly(ethyleneglycol) diacrylate (PEGDA) M(n)∼258 as the single monomer, a mixture of
B Thanuja et al.
Ultrasonics sonochemistry, 19(6), 1213-1220 (2012-04-14)
Density (ρ), ultrasonic velocity (U), for the binary mixtures of 4-methoxy benzoin (4MB) with ethanol, chloroform, acetonitrile, benzene, and di-oxane were measured at 298K. The solute-solvent interactions and the effect of the polarity of the solvent on the type of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service