Skip to Content
Merck
All Photos(1)

Key Documents

610046

Sigma-Aldrich

Boron-11B

95 atom % 11B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
11B
CAS Number:
Molecular Weight:
11.01
MDL number:
UNSPSC Code:
12141907
PubChem Substance ID:

isotopic purity

95 atom % 11B

form

solid

mass shift

depleted

SMILES string

[11B]

InChI

1S/B/i1+0

InChI key

ZOXJGFHDIHLPTG-IGMARMGPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chathura D Suraweera et al.
Cell death & disease, 9(10), 967-967 (2018-09-22)
Bcl-2 family proteins play a crucial role in regulating apoptosis, a process critical for development, eliminating damaged or infected cells, host-pathogen interactions and in disease. Dysregulation of Bcl-2 proteins elicits an expansive cell survival mechanism promoting cell migration, invasion and
John C Rose et al.
Nature methods, 14(9), 891-896 (2017-07-25)
We developed a chemically inducible Cas9 (ciCas9) and a droplet digital PCR assay for double-strand breaks (DSB-ddPCR) to investigate the kinetics of Cas9-mediated generation and repair of DSBs in cells. ciCas9 is a rapidly activated, single-component Cas9 variant engineered by
Michael Fricker et al.
Physiological reviews, 98(2), 813-880 (2018-03-01)
Neuronal cell death occurs extensively during development and pathology, where it is especially important because of the limited capacity of adult neurons to proliferate or be replaced. The concept of cell death used to be simple as there were just
J Zhang et al.
Cell death & disease, 7, e2266-e2266 (2016-06-17)
How BH3-only proteins activate Bax/Bak, the two gateway proteins of the mitochondria-dependent apoptotic pathway, remains incompletely understood. Although all pro-apoptotic BH3-only proteins are known to bind/neutralize the anti-apoptotic Bcl-2 proteins, the three most potent ones, Bid (tBid), Bim, and Puma
S Matt Gilbert et al.
Scientific reports, 7(1), 15096-15096 (2017-11-10)
We demonstrate the fabrication of individual nanopores in hexagonal boron nitride (h-BN) with atomically precise control of the pore shape and size. Previous methods of pore production in other 2D materials typically create pores with irregular geometry and imprecise diameters.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service