Skip to Content
Merck
All Photos(1)

Key Documents

606154

Sigma-Aldrich

Vanillin-(methoxy-13C)

99 atom % 13C

Synonym(s):

4-Hydroxy-3-methoxy-13C-benzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC7H8O3
CAS Number:
Molecular Weight:
153.14
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:

isotopic purity

99 atom % 13C

form

solid

technique(s)

mass spectrometry (MS): suitable

bp

170 °C/15 mmHg (lit.)

mp

81-83 °C (lit.)

mass shift

M+1

SMILES string

[13CH3]Oc1cc(C=O)ccc1O

InChI

1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3/i1+1

InChI key

MWOOGOJBHIARFG-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

319.6 - 321.4 °F - closed cup

Flash Point(C)

159.8 - 160.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service