Skip to Content
Merck
All Photos(1)

Documents

602647

Sigma-Aldrich

Ritalinic acid

99% (CP)

Synonym(s):

a-Phenyl-2-piperidineacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO2
CAS Number:
Molecular Weight:
219.28
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

Assay

99% (CP)

SMILES string

OC(=O)C(C1CCCCN1)c2ccccc2

InChI

1S/C13H17NO2/c15-13(16)12(10-6-2-1-3-7-10)11-8-4-5-9-14-11/h1-3,6-7,11-12,14H,4-5,8-9H2,(H,15,16)

InChI key

INGSNVSERUZOAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Aoyama et al.
Journal of pharmaceutical sciences, 79(6), 465-469 (1990-06-01)
A gas chromatographic-mass spectrometric method was developed for the stereoselective quantification of threo-methylphenidate (MPD) and its metabolite, ritalinic acid (RA), in plasma or urine. The plasma concentrations of (+)-MPD after oral administration of two 10-mg conventional tablets containing racemic MPD.HCl
Marion Letzel et al.
Chemosphere, 81(11), 1416-1422 (2010-10-12)
To investigate the occurrence and fate of ritalinic acid - the main human metabolite of the psychostimulant drug methylphenidate - in the aquatic environment, a HPLC-electrospray-MS/MS method for the quantification of ritalinic acid in wastewater, surface water and bank filtrate
A Solans et al.
Journal of chromatography. B, Biomedical applications, 658(2), 380-384 (1994-08-19)
Two analytical methods for the simultaneous detection in urine of methylphenidate and its main metabolite, ritalinic acid, are described. Both procedures are based on solid-phase extraction of urine samples on Bond Elut Certify columns, and capillary gas chromatographic-mass spectrometric detection
N R Srinivas et al.
Journal of chromatography, 530(2), 327-336 (1990-09-14)
Enantioselective gas chromatographic assays for the quantitation of methylphenidate and its major metabolite ritalinic acid in plasma are described. The procedures involved the extraction of methylphenidate enantiomers from alkanised plasma. The plasma was then washed to ensure complete removal of
D R Doerge et al.
Rapid communications in mass spectrometry : RCM, 14(8), 619-623 (2000-04-29)
Methylphenidate (MP, Ritalin) is a psychotropic drug widely prescribed to children for treating the symptoms of attention deficit disorder with and without hyperactivity. Because little information exists about the effects of chronic MP administration on cognitive function in children, measures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service