Skip to Content
Merck
All Photos(1)

Documents

594768

Sigma-Aldrich

4-Methoxycarbonylphenylboronic acid pinacol ester

97%

Synonym(s):

4-Carbomethoxyphenylboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19BO4
CAS Number:
Molecular Weight:
262.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

77-81 °C (lit.)

SMILES string

COC(=O)c1ccc(cc1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C14H19BO4/c1-13(2)14(3,4)19-15(18-13)11-8-6-10(7-9-11)12(16)17-5/h6-9H,1-5H3

InChI key

REIZEQZILPXYKS-UHFFFAOYSA-N

Application

4-Methoxycarbonylphenylboronic acid pinacol ester can be used as a reagent:   
  • In Suzuki–Miyaura cross-coupling reaction with aryl halides to form C-C bonds.     
  • For the synthesis of biphenyl derivatives by selective ortho C-H arylation of ketones using Rh catalyst.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelvin S L Chan et al.
Nature chemistry, 6(2), 146-150 (2014-01-24)
There have been numerous developments in C-H activation reactions in the past decade. Attracted by the ability to functionalize molecules directly at ostensibly unreactive C-H bonds, chemists have discovered reaction conditions that enable reactions of C(sp(2))-H and C(sp(3))-H bonds with
Bing Zhang et al.
Organic letters, 19(21), 5940-5943 (2017-10-20)
A general method for selective ortho C-H arylation of ketone, with boron reagent enabled by rhodium complexes with excellent yields, is developed. The transformation is characterized by the use of air-stable Rh catalyst, high monoarylation selectivity, and excellent yields of
Synthesis of ester-substituted dihydroacridine derivatives and their spectroscopic properties
Suzuki R, et al.
New. J. Chem., 40(3), 2920-2926 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service