Skip to Content
Merck
All Photos(1)

Documents

589756

Sigma-Aldrich

Tetradecanoic-14,14,14-d3 acid

99 atom % D, 98% (CP)

Synonym(s):

Myristic acid-14,14,14-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CH2)12CO2H
CAS Number:
Molecular Weight:
231.39
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % D

Quality Level

Assay

98% (CP)

form

solid

bp

250 °C/100 mmHg (lit.)

mp

52-54 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])CCCCCCCCCCCCC(O)=O

InChI

1S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)/i1D3

InChI key

TUNFSRHWOTWDNC-FIBGUPNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhanru Yu et al.
Talanta, 165, 685-691 (2017-02-06)
Two-dimensional gas chromatography mass spectrometry (GCxGC-MS) is utilized to an increasing extent in biomedical metabolomics. Here, we established and adapted metabolite extraction and derivatization protocols for cell/tissue biopsy, serum and urine samples according to their individual properties. GCxGC-MS analysis revealed
Zhanru Yu et al.
Talanta, 199, 184-188 (2019-04-07)
Polyamines are a class of poly-cationic aliphatic amines, playing a role in different cellular processes such as maintaining intracellular pH and membrane potential that are relevant for general cellular physiology and ageing. The development of analytical methods for detection and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service