589586
Propionic acid-13C3
99 atom % 13C
Synonym(s):
Propanoic acid-13C3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
13CH313CH213CO2H
CAS Number:
Molecular Weight:
77.06
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
99 atom % 13C
Assay
99% (CP)
bp
141 °C (lit.)
mp
-24--23 °C (lit.)
density
1.024 g/mL at 25 °C
mass shift
M+3
SMILES string
[13CH3][13CH2][13C](O)=O
InChI
1S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)/i1+1,2+1,3+1
InChI key
XBDQKXXYIPTUBI-VMIGTVKRSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
129.2 °F - closed cup
Flash Point(C)
54 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Stacy A Voils et al.
Thrombosis research, 165, 6-13 (2018-03-16)
Incidence of venous thromboembolism (VTE) in critically ill patients remains unacceptably high despite widespread use of thromboprophylaxis. A systems biology approach may be useful in understanding disease pathology and predicting response to treatment. Metabolite profile under specific environmental conditions provides
Madeleine Wyss et al.
Frontiers in immunology, 10, 3107-3107 (2020-02-06)
Early life exposure to microbes plays an important role in immune system development. Germ-free mice, or mice colonized with a low-diversity microbiota, exhibit high serum IgE levels. An increase in microbial richness, providing it occurs in a critical developmental window
Adam Shute et al.
Microbiome, 9(1), 186-186 (2021-09-15)
Studies on the inhibition of inflammation by infection with helminth parasites have, until recently, overlooked a key determinant of health: the gut microbiota. Infection with helminths evokes changes in the composition of their host's microbiota: one outcome of which is
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service