Skip to Content
Merck
All Photos(1)

Documents

586765

Sigma-Aldrich

Aniline-4-13C

99 atom % 13C

Synonym(s):

Benzenamine-4-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CC5H4NH2
CAS Number:
Molecular Weight:
93.11
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C

bp

184 °C (lit.)

mp

-6 °C (lit.)

density

1.033 g/mL at 25 °C

mass shift

M+1

storage temp.

2-8°C

SMILES string

Nc1cc[13cH]cc1

InChI

1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2/i1+1

InChI key

PAYRUJLWNCNPSJ-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Resp. Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timo Stahl et al.
Journal of the American Chemical Society, 135(4), 1248-1251 (2013-01-15)
Heterolytic splitting of the Si-H bond mediated by a Ru-S bond forms a sulfur-stabilized silicon cation that is sufficiently electrophilic to abstract fluoride from CF(3) groups attached to selected anilines. The ability of the Ru-H complex, generated in the cooperative
Chia-Chi Su et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 48(9), 1012-1018 (2013-04-12)
This study was undertaken to investigate the feasibility of applying the Fered-Fenton process to the degradation of m-phenylenediamine, by examining the effect of varying the initial H2O2 and Fe(2+) concentrations, the initial pH and electric current on the process efficiency.
Regina Mažeikienė et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 106, 34-40 (2013-01-30)
For the first time, chemical oxidation of aniline and N-methylaniline with dichromate as oxidant has been studied by Raman spectroscopy with 785 nm laser beam excitation, and the suitability of this technique for kinetic study of this process was demonstrated.
Ronald Besandre et al.
Organic letters, 15(7), 1666-1669 (2013-03-15)
N-substituted indoles are synthesized from primary amines through a tandem reaction sequence. Initial condensation of the amine with an α-(o-haloaryl)ketone or aldehyde is followed by intramolecular aryl amination catalyzed by CuI. A variety of anilines and alkyl amines, including those
Yu Qian et al.
Chemical communications (Cambridge, England), 49(26), 2700-2702 (2013-02-26)
An enantioselective four-component reaction of a diazoketone, water, an aniline and ethyl glyoxylate in the presence of catalytic Rh2(OAc)4 and a chiral Brønsted acid was developed to efficiently produce β-hydroxy-α-amino acid derivatives in good yields with high diastereoselectivity and enantioselectivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service