Skip to Content
Merck
All Photos(2)

Key Documents

566268

Sigma-Aldrich

Barium iodide dihydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bal2 · 2H2O
CAS Number:
Molecular Weight:
427.17
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
form:
solid

Assay

98%

form

solid

density

5.15 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].I[Ba]I

InChI

1S/Ba.2HI.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LRUCFPBOVNFNMO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Barium iodide dihydrate can be used as a barium precursor for the preparation of barium iodo alkoxide/acetato complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Redistribution reactions of heteroleptic barium iodide derivatives: Synthesis and structures of trans-BaI2 (DME)(triglyme), cis-BaI2 (DME)(tetraglyme) and [Ba (tetraglyme) 2] I2
Mishra S, et al.
Polyhedron, 26(1), 66-72 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service