563757
2,3-Dimethylphenylmagnesium bromide
0.5 M in THF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2C6H3MgBr
CAS Number:
Molecular Weight:
209.37
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
reaction suitability
reaction type: Grignard Reaction
concentration
0.5 M in THF
bp
65 °C (lit.)
density
0.948 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
Cc1cccc([Mg]Br)c1C
InChI
1S/C8H9.BrH.Mg/c1-7-5-3-4-6-8(7)2;;/h3-5H,1-2H3;1H;/q;;+1/p-1
InChI key
GWICKWUOSJJCNO-UHFFFAOYSA-M
Related Categories
Legal Information
Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service