Skip to Content
Merck
All Photos(2)

Key Documents

560979

Sigma-Aldrich

Silica, mesostructured

MSU-F (cellular foam)

Synonym(s):

Silica, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder

cell window size

~15 nm

surface area

562 m2/g

unit cell size

~22 nm

pore size

2.31 cm3/g pore volume

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David M Brown et al.
Toxicology letters, 224(1), 147-156 (2014-01-28)
We investigated the effects of silica particles and nanoparticles (NPs) (50 nm and 200 nm) with a neutral and positively charged surface when dispersed in saline, bovine serum albumin (BSA) or lung lining fluid (LLF) 24 h post instillation into
Richard Hayes et al.
Journal of chromatography. A, 1357, 36-52 (2014-05-27)
The challenges in HPLC are fast and efficient separation for a wide range of samples. Fast separation often results in very high operating pressure, which places a huge burden on HPLC instrumentation. In recent years, core-shell silica microspheres (with a
Fabrice Gritti et al.
Journal of chromatography. A, 1342, 24-29 (2014-04-17)
The advantages of using columns packed with stationary phases having a composition gradient so that retention factors increase toward the outlet and eluting them with an isocratic mobile phase may provide increased peak resolution in liquid chromatography. This approach is
Yaswanth Kuthati et al.
Journal of nanoscience and nanotechnology, 13(4), 2399-2430 (2013-06-15)
The advent of Nanotechnology has paved a way for improved disease treatment strategies, the most noteworthy being the mesoporous silica nanoparticles (MSNs) which have gained much recent attention in the field of cancer therapy and its diagnosis. The flaws of
Graham J Edgar et al.
Nature, 506(7487), 216-220 (2014-02-07)
In line with global targets agreed under the Convention on Biological Diversity, the number of marine protected areas (MPAs) is increasing rapidly, yet socio-economic benefits generated by MPAs remain difficult to predict and under debate. MPAs often fail to reach

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service