Skip to Content
Merck
All Photos(2)

Key Documents

559814

Sigma-Aldrich

1-Naphthyl-N-methylcarbamate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OCONHCH3
CAS Number:
Molecular Weight:
201.22
Beilstein:
1875862
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

142-146 °C (lit.)

SMILES string

CNC(=O)Oc1cccc2ccccc12

InChI

1S/C12H11NO2/c1-13-12(14)15-11-8-4-6-9-5-2-3-7-10(9)11/h2-8H,1H3,(H,13,14)

InChI key

CVXBEEMKQHEXEN-UHFFFAOYSA-N

Gene Information

rat ... Faah(29347)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michele M Schantz et al.
Analytical and bioanalytical chemistry, 407(11), 2945-2954 (2015-02-06)
Two new Standard Reference Materials (SRMs), SRM 3672 Organic Contaminants in Smokers' Urine (Frozen) and SRM 3673 Organic Contaminants in Non-Smokers' Urine (Frozen), have been developed in support of studies for assessment of human exposure to select organic environmental contaminants.
Carbaryl. A toxicological review and risk analysis.
M F Cranmer
Neurotoxicology, 7(1), 247-328 (1986-01-01)
Amrith S Gunasekara et al.
Reviews of environmental contamination and toxicology, 196, 95-121 (2008-11-26)
Carbaryl is an agricultural and garden insecticide that controls a broad spectrum of insects. Although moderately water soluble, it neither vaporizes nor volatilizes readily. However, upon spray application the insecticide is susceptible to drift. It is unstable under alkaline conditions
Dieter I M De Coninck et al.
Aquatic toxicology (Amsterdam, Netherlands), 130-131, 149-159 (2013-02-16)
Natural and chemical stressors occur simultaneously in the aquatic environment. Their combined effects on biota are usually difficult to predict from their individual effects due to interactions between the different stressors. Several recent studies have suggested that synergistic effects of
Jordan L Burke et al.
Pest management science, 68(7), 1048-1052 (2012-02-24)
Tests were conducted on two insecticides (carbaryl and bifenthrin) for excluding subcortical beetles (Coleoptera: Curculionidae and Cerambycidae) from loblolly pine trees (Pinus taeda L.). Two trap designs (single- and double-pane windows) and two trapping heights (1.5 and 4 m) were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service