Skip to Content
Merck
All Photos(1)

Key Documents

55875

Sigma-Aldrich

2-Hydroxy-4-(methylthio)butyric acid calcium salt

≥99.0% (NT)

Synonym(s):

Calcium 2-hydroxy-4-(methylthio)butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3SCH2CH2CH(OH)COO]2Ca
CAS Number:
Molecular Weight:
338.45
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (NT)

form

solid

functional group

hydroxyl
thioether

storage temp.

2-8°C

SMILES string

CSCCC(O)C(=O)O[Ca]OC(=O)C(O)CCSC

InChI

1S/2C5H10O3S.Ca/c2*1-9-3-2-4(6)5(7)8;/h2*4,6H,2-3H2,1H3,(H,7,8);/q;;+2/p-2

InChI key

ABRVDWASZFDIEH-UHFFFAOYSA-L

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sheng-Yao Kuang et al.
Fish & shellfish immunology, 32(5), 629-636 (2012-01-17)
Immune response and antioxidant status of immune organs in juvenile Jian carp (Cyprinus carpio var. Jian) fed graded levels of methionine hydroxy analogue (MHA) (0, 5.1, 7.6, 10.2, 12.7, 15.3 g kg(-1) diet) for 60 days were investigated. Results indicated
Z F Fang et al.
Amino acids, 36(3), 501-509 (2008-06-06)
To determine whether portal plasma flow (PPF) and net portal appearance of amino acids (AA) could be affected by 2-hydroxy-4-methylthiobutyrate (HMB), six barrows (35-day-old, 8.6+/-1.4 kg), implanted with arterial, portal and mesenteric catheters, were fed a DL-methionine (as the control)
H Willemsen et al.
Poultry science, 90(10), 2311-2320 (2011-09-22)
Heat stress is known to impair performance and to induce oxidative stress in poultry. The aim of the present study was to compare the effects of dietary supplementation of dl-methionine (dl-M) or the synthetic analog 2-hydroxy-4-methylthiobutanoic acid (dl-HMTBA) on broiler
A Vieira-Neto et al.
Journal of dairy science, 104(1), 683-701 (2020-11-10)
Objectives were to determine the effects of an injectable formulation of calcitriol on Ca concentration, risk of clinical diseases, and performance in dairy cows. Cows were blocked by lactation number (1 vs. >1) and calving sequence and, within block, assigned
R Martín-Venegas et al.
Poultry science, 87(7), 1392-1394 (2008-06-26)
Methionine hydroxy analogue DL-2-hydroxy-(4-methylthio)butanoic acid (HMTBA) is commonly used as a supplemental source of Met in commercial animal diets. To better understand the uptake of this analogue by the chicken intestine, the aim of the present study was to assess

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service