Skip to Content
Merck
All Photos(2)

Documents

557927

Sigma-Aldrich

1-Ethynylnaphthalene

97%

Synonym(s):

α-Ethynylnaphthalene, α-Naphthylacetylene, 1-Ethynylnaphthalene, 1-Naphthylacetylene, 1-Naphthylethyne

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7C≡CH
CAS Number:
Molecular Weight:
152.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.6500 (lit.)

density

1.070 g/mL at 25 °C (lit.)

SMILES string

C#Cc1cccc2ccccc12

InChI

1S/C12H8/c1-2-10-7-5-8-11-6-3-4-9-12(10)11/h1,3-9H

InChI key

MCZUXEWWARACSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethynylnaphthalene can be prepared from 1-acetonaphthone or 1-naphthyl-2-trimethylsilylacetylene.

Application

1-Ethynylnaphthalene may be used to synthesize bis-6,6′-(ethynyl-1-naphthalene)-2,2′-bipyridine.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.0 °F - closed cup

Flash Point(C)

57.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G J Hammons et al.
Chemical research in toxicology, 2(6), 367-374 (1989-11-01)
Since the N-oxidation of several carcinogenic arylamines has been shown to be catalyzed preferentially by cytochrome P-450IA2 in several species, homologous ethynyl-substituted aromatic hydrocarbons, 2-ethynylnaphthalene, 1-ethynylnaphthalene, and 2-ethynylfluorene, were synthesized and examined as potential mechanism-based inactivators of this monooxygenase. By

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service