Skip to Content
Merck
All Photos(1)

Key Documents

551295

Sigma-Aldrich

N-Cyclopropyl-2,4,6-triamino-1,3,5-triazine

97%

Synonym(s):

N-Cyclopropyl-1,3,5-triazin-2,4,6-triamine, Cyromazin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N6
CAS Number:
Molecular Weight:
166.18
Beilstein:
882879
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

223-227 °C (lit.)

SMILES string

Nc1nc(N)nc(NC2CC2)n1

InChI

1S/C6H10N6/c7-4-10-5(8)12-6(11-4)9-3-1-2-3/h3H,1-2H2,(H5,7,8,9,10,11,12)

InChI key

LVQDKIWDGQRHTE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Cyclopropyl-2,4,6-triamino-1,3,5-triazine, also known as cyromazine, is an insect growth regulator(IGR) that is commonly used as an insecticide. It undergoes metabolism in plants and animals to form melamine. Surface-enhanced Raman scattering (SERS) spectroscopic study indicates that it interacts with the surface of silver colloid through the lone pair on nitrogen.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raman and surface-enhanced Raman study of insecticide cyromazine.
Mukherjee K, et al.
Vibrational Spectroscopy, 25(1), 91-99 (2001)
G W Levot
Australian veterinary journal, 90(11), 433-437 (2012-10-31)
To investigate the cause of cyromazine failure to protect lambs from flystrike. Lucilia cuprina larvae from a Nimmitabel (New South Wales) population associated with failure of a cyromazine spray-on to protect lambs from flystrike were compared with larvae from a
Xuegang Li et al.
Journal of separation science, 34(3), 323-330 (2011-01-27)
In this study, we described a high-sensitive on-line preconcentration method for cypromazine (CYP) and melamine (MEL) analysis using cation-selective exhaustive injection (CSEI) combined with sweeping-MEKC. The optimum conditions of on-line concentration and separation were discussed. The BGE contained 100 mM SDS
Shanshan Wang et al.
The Analyst, 136(18), 3672-3679 (2011-07-26)
We report a novel method for simultaneous determination of cyromazine and melamine based on a molecularly imprinted monolith on-line coupled with high performance liquid chromatography (HPLC). The imprinted monolith was prepared by in situ polymerization using 2,4-diamino-6-undecyl-1,3,5-triazine (DAUTA) as a
Xianhua Wang et al.
Journal of separation science, 35(12), 1432-1438 (2012-06-29)
The combination of molecularly imprinted polymer with high performance liquid chromatography has been developed to determine cyromazine and its metabolic melamine in some samples. However, the potential risk of template leakage used in molecularly imprinted polymer is a major disadvantage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service