Skip to Content
Merck
All Photos(1)

Key Documents

549053

Sigma-Aldrich

3-Bromothioanisole

97%

Synonym(s):

2-Bromophenyl methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4SCH3
CAS Number:
Molecular Weight:
203.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.628 (lit.)

bp

124-125 °C/10 mmHg (lit.)

density

1.51 g/mL at 25 °C (lit.)

functional group

bromo
thioether

SMILES string

CSc1cccc(Br)c1

InChI

1S/C7H7BrS/c1-9-7-4-2-3-6(8)5-7/h2-5H,1H3

InChI key

NKYFJZAKUPSUSH-UHFFFAOYSA-N

General description

3-Bromothioanisole is a 3-halothioanisole derivative that is mainly used in the preparation of photoacids. It can be prepared from 3-bromobenzenethiol.

Application

3-Bromothioanisole may be used in the preparation of:
  • 3-methylthiotriphenylamine
  • 4-ethoxy-3′-methylthiostilbene
  • 3-bromophenyl phenyl sulfide
  • 9-substituted, 3,6-dithiomethylfluorenes
  • 4-methoxy-3′-(methylthio)-1,1′-biphenyl

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transition-Metal-Free Acid-Mediated Synthesis of Aryl Sulfides from Thiols and Thioethers.
Wagner AM and Sanford MS.
The Journal of Organic Chemistry, 79(5), 2263-2267 (2014)
A Preparatively Convenient Ligand-Free Catalytic PEG 2000 Suzuki- Miyaura Coupling.
Razler TM, et al.
The Journal of Organic Chemistry, 74(3), 1381-1384 (2008)
Enhancement of acid photogeneration through a para-to-meta substitution strategy in a sulfonium-based alkoxystilbene designed for two-photon polymerization.
Xia R, et al.
Chemistry of Materials, 24(2), 237-244 (2012)
Efficient photoacids based upon triarylamine dialkylsulfonium salts.
Zhou W, et al.
Journal of the American Chemical Society, 124(9), 1897-1901 (2002)
Evaluating atomic components in fluorene wires.
Klausen RS, et al.
Chemical Science, 5(4), 1561-1564 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service