Skip to Content
Merck
All Photos(1)

Key Documents

544051

Sigma-Aldrich

L-Homophenylalanine hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2CH(NH2)CO2H · HCl
CAS Number:
Molecular Weight:
215.68
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

optical activity

[α]20/D +38°, c = 1 in 3 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

mp

262-265 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Cl.N[C@@H](CCc1ccccc1)C(O)=O

InChI

1S/C10H13NO2.ClH/c11-9(10(12)13)7-6-8-4-2-1-3-5-8;/h1-5,9H,6-7,11H2,(H,12,13);1H/t9-;/m0./s1

InChI key

CHBMONIBOQCTCF-FVGYRXGTSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tasir S Haque et al.
Peptides, 31(5), 950-955 (2010-02-09)
We report the identification of potent agonists of the Glucagon-Like Peptide-1 Receptor (GLP-1R). These compounds are short, 11 amino acid peptides containing several unnatural amino acids, including (in particular) analogs of homohomophenylalanine (hhPhe) at the C-terminal position. Typically the functional
A L Ahmad et al.
Biotechnology advances, 27(3), 286-296 (2009-06-09)
Over the past decade, L-homophenylalanine is extensively used in the pharmaceutical industry as a precursor for production of angiotensin-converting enzyme (ACE) inhibitor, which possesses significant clinical application in the management of hypertension and congestive heart failure (CHF). A number of
Adeyemi O Aremu et al.
Journal of plant physiology, 169(15), 1530-1541 (2012-08-14)
The effect of five topolins (meta-Topolin=mT; meta-Topolin riboside=mTR; meta-Methoxy topolin=MemT; meta-Methoxy topolin riboside=MemTR and 6-(meta-methoxy)-9-(tetrahydropyran-2-yl)-topolin=MemTTHP) on the photosynthetic pigments and leaf structures of micropropagated 'Williams' bananas was compared with the commonly used benzyladenine (BA). Surface-decontaminated explants were cultured for 70
Sonja Nordhoff et al.
Bioorganic & medicinal chemistry letters, 19(15), 4201-4203 (2009-06-12)
Modifications of DPP-4 inhibitor 5, that was discovered by structure based design, are described and structure-activity relationships discussed. With analogue 7k one of the most potent non-covalent inhibitors of DPP-4 reported to date (IC(50)=0.38nM) was discovered. X-ray structure of inhibitor
Candela Cuesta et al.
Journal of plant physiology, 166(11), 1162-1171 (2009-02-24)
Adventitious bud formation in stone pine cotyledons cultured in the presence of benzyladenine (BA) has been proposed as a model for the study of in vitro shoot organogenesis in conifers. This is because of its advantageous characteristics including the requirement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service